2-Methylthiophene-3-carbonyl chloride 90%
Dodavatel: Apollo Scientific
APOSOR908212-1GEA
15340
CZK
APOSOR908212-1G
2-Methylthiophene-3-carbonyl chloride 90%
2-Methylthiophene-3-carbonyl chloride
Vzorec:
C₆H₅ClOS MW: 160,621 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18417771 CAS číslo: 123116-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...