4,4,8-Trimethyl-2,3-dihydro-1H-quinoline 95%
Dodavatel: Apollo Scientific
APOSOR908225-1GEA
16370
CZK
APOSOR908225-1G
4,4,8-Trimethyl-2,3-dihydro-1H-quinoline 95%
4,4,8-Trimethyl-2,3-dihydro-1H-quinoline
Vzorec:
C₁₂H₁₇N MW: 175,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12913836 CAS číslo: 1187933-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...