7-Chloro-4,4-dimethyl-2,3-dihydro-1H-quinoline 95%
Dodavatel: Apollo Scientific
APOSOR908251-1GEA
24630
CZK
APOSOR908251-1G
7-Chloro-4,4-dimethyl-2,3-dihydro-1H-quinoline 95%
7-Chloro-4,4-dimethyl-2,3-dihydro-1H-quinoline
Vzorec:
C₁₁H₁₄ClN MW: 195,689 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12913832 CAS číslo: 1187933-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...