2,4,6-trimethylbenzenesulphinic acid 96%
Dodavatel: Apollo Scientific
APOSOR908265-1GEA
18020
CZK
APOSOR908265-1G
2,4,6-trimethylbenzenesulphinic acid 96%
2,4,6-trimethylbenzenesulphinic acid
Vzorec:
C₉H₁₂O₂S MW: 184,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03940230 CAS číslo: 59057-35-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...