2-Cyano-1,3-dihydroindene-2-carboxylic acid 97%
Dodavatel: Apollo Scientific
APOSOR909657-1GEA
9290
CZK
APOSOR909657-1G
2-Cyano-1,3-dihydroindene-2-carboxylic acid 97%
2-Cyano-1,3-dihydroindene-2-carboxylic acid
Vzorec:
C₁₁H₉NO₂ MW: 187,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21802888 CAS číslo: 858226-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...