Tisk…

3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile 98%

Dodavatel: Apollo Scientific

APOSOR910325-1GEA 9130 CZK
APOSOR910325-1G
3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile 98%
3-(5-Hydroxy-3-methylpyrazol-1-yl)propanenitrile
Vzorec: C₇H₉N₃O
MW: 151,166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00634617
CAS číslo: 41534-08-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR