Tisk…

Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate 96%

Dodavatel: Apollo Scientific

APOSOR910353-1GEA 14290 CZK
APOSOR910353-1G
Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate 96%
Methyl 4,5,6,7-tetrahydro-1H-benzimidazole-5-carboxylate
Vzorec: C₉H₁₂N₂O₂
MW: 180,204 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20662345
CAS číslo: 1437794-30-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR