Tisk…

1-Methanesulphonylindoline 98%

Dodavatel: Apollo Scientific

APOSOR910577-5GEA 28570 CZK
APOSOR910577-5G APOSOR910577-1G
1-Methanesulphonylindoline 98%
1-Methanesulphonylindoline
Vzorec: C₉H₁₁NO₂S
MW: 197,254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01213175
CAS číslo: 5825-63-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR