Tisk…

3-cyano-4-ethoxybenzoic acid 95%

Dodavatel: Apollo Scientific

APOSOR911191-1GEA 18520 CZK
APOSOR911191-1G
3-cyano-4-ethoxybenzoic acid 95%
3-cyano-4-ethoxybenzoic acid
Vzorec: C₁₀H₉NO₃
MW: 191,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09271828
CAS číslo: 258273-32-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR