Tisk…

3-Chloro-1H-quinolin-2-one 95%

Dodavatel: Apollo Scientific

APOSOR911269-1GEA 15340 CZK
APOSOR911269-1G
3-Chloro-1H-quinolin-2-one 95%
3-Chloro-1H-quinolin-2-one
Vzorec: C₉H₆ClNO
MW: 179,603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705671
CAS číslo: 939-17-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR