Tisk…

4-Chloro-3-(cyclopropylmethoxy)aniline 95%

Dodavatel: Apollo Scientific

APOSOR912358-1GEA 18020 CZK
APOSOR912358-1G
4-Chloro-3-(cyclopropylmethoxy)aniline 95%
4-Chloro-3-(cyclopropylmethoxy)aniline
Vzorec: C₁₀H₁₂ClNO
MW: 197,661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19625029
CAS číslo: 1265236-35-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR