Tisk…

6-Ethyl-2-naphthalenol 95%

Dodavatel: Apollo Scientific

APOSOR916439-1GEA 4930 CZK
APOSOR916439-1G
6-Ethyl-2-naphthalenol 95%
6-Ethyl-2-naphthalenol
Vzorec: C₁₂H₁₂O
MW: 172,223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368645
CAS číslo: 1999-64-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR