Tisk…

(3,5-Dichloropyridin-4-yl)methanamine 95%

Dodavatel: Apollo Scientific

APOSOR916476-1GEA 20500 CZK
APOSOR916476-1G
(3,5-Dichloropyridin-4-yl)methanamine 95%
(3,5-Dichloropyridin-4-yl)methanamine
Vzorec: C₆H₆Cl₂N₂
MW: 177,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18254248
CAS číslo: 418795-04-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR