5-Phenyl-pyrimidine 95%
Dodavatel: Apollo Scientific
APOSOR916707-1GEA
22980
CZK
APOSOR916707-1G
5-Phenyl-pyrimidine 95%
5-Phenyl-pyrimidine
Vzorec:
C₁₀H₈N₂ MW: 156,184 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234616 CAS číslo: 34771-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...