Tisk…

6-Methyl-1H-pyrazolo[3,4-b]pyridine 95%

Dodavatel: Apollo Scientific

APOSOR917539-1GEA 5690 CZK
APOSOR917539-1G
6-Methyl-1H-pyrazolo[3,4-b]pyridine 95%
6-Methyl-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₇H₇N₃
MW: 133,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234559
CAS číslo: 885269-66-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR