1-Methyl-4-phenoxybenzene 95%
Dodavatel: Apollo Scientific
APOSOR917922-1GEA
2530
CZK
APOSOR917922-1G
1-Methyl-4-phenoxybenzene 95%
1-Methyl-4-phenoxybenzene
Vzorec:
C₁₃H₁₂O MW: 184,234 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043948 CAS číslo: 1706-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...