2-tert-Butyl-1,3,4-oxadiazole 95%
Dodavatel: Apollo Scientific
APOSOR918655-1GEA
18450
CZK
APOSOR918655-1G
2-tert-Butyl-1,3,4-oxadiazole 95%
2-tert-Butyl-1,3,4-oxadiazole
Vzorec:
C₆H₁₀N₂O MW: 126,156 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08689904 CAS číslo: 251540-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...