Tisk…

5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95+%

Dodavatel: Apollo Scientific

Varování

APOSOR919134-250MGEA 5290 CZK
APOSOR919134-250MG
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95+%
5-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one
Vzorec: C₇H₅BrN₂O
MW: 213,031 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848386
CAS číslo: 1217002-90-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR