Methyl 3-amino-2,4-dimethylbenzoate 95%
Dodavatel: Apollo Scientific
APOSOR919396-1GEA
6050
CZK
APOSOR919396-1G
Methyl 3-amino-2,4-dimethylbenzoate 95%
Methyl 3-amino-2,4-dimethylbenzoate
Vzorec:
C₁₀H₁₃NO₂ MW: 179,216 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22494937 CAS číslo: 24812-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...