Tisk…

Ethyl 4-formamidobenzoate 95%

Dodavatel: Apollo Scientific

APOSOR921120-1GEA 1500 CZK
APOSOR921120-1G
Ethyl 4-formamidobenzoate 95%
Ethyl 4-formamidobenzoate
Vzorec: C₁₀H₁₁NO₃
MW: 193,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00021025
CAS číslo: 5422-63-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR