Order Entry
2-Methyl-4-(trimethylsilyl)but-3-yn-2-ol 95%
2-Methyl-4-(trimethylsilyl)but-3-yn-2-ol 95%
Supplier:  Apollo Scientific
Certificates
2-Methyl-4-(trimethylsilyl)but-3-yn-2-ol 95%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSOR942138-1G
    OR942138-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 612,00 Kč
    612,00 Kč
    1 * 1 g
       612,00 Kč
    Supplier Article Number: OR942138-1G
    Local Article Number: APOSOR942138-1G
  • APOSOR942138-5G
    OR942138-5G
    Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 655,00 Kč
    655,00 Kč
    1 * 5 g
       655,00 Kč
    Supplier Article Number: OR942138-5G
    Local Article Number: APOSOR942138-5G