Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Czech Republic
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 Kč
Order Entry
Home
2,5-Dibromo-4-(trifluoromethoxy)aniline
2,5-Dibromo-4-(trifluoromethoxy)aniline 98%
2,5-Dibromo-4-(trifluoromethoxy)aniline 98%
2,5-Dibromo-4-(trifluoromethoxy)aniline 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2,5-Dibromo-4-(trifluoromethoxy)aniline 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
84483-32-9
:
Formula:
C₇H₄Br₂F₃NO
MDL Number:
MFCD28785127
:
334.916 g/mol
Storage Temperature:
Ambient
Product Information
Catalog #
Supplier #
Pack type
Pk
Availability
Price
APOSPC303351-1G
PC303351-1G
Bottle
1 g
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 7 080,00 Kč
7 080,00 Kč
-
+
Add to cart
1 * 1 g
7 080,00 Kč
-
+
Add to cart
APOSPC303351-5G
PC303351-5G
Bottle
5 g
Log in
for availability
Log in
for availability
1 * 5 g
1 * 5 g - 24 670,00 Kč
24 670,00 Kč
-
+
Add to cart
1 * 5 g
24 670,00 Kč
-
+
Add to cart