Potassium (5-fluoro-2-formylphenyl)trifluoroborate
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₇H₄BF₄O·K
MDL Number:MFCD28963754
:230.009 g/mol
:GHS07
Storage Temperature:Ambient
Product Information
- Catalog #Supplier #Pack typePkAvailabilityPrice
- Supplier Article Number: PC53071-1GLocal Article Number: APOSPC53071-1G
- Supplier Article Number: PC53071-5GLocal Article Number: APOSPC53071-5G