Order Entry
2-(2,6-Difluoro-3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
2-(2,6-Difluoro-3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier:  Apollo Scientific
Certificates
2-(2,6-Difluoro-3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97%
Supplier:  Apollo Scientific
Product Information
  • Catalog #
    Supplier #
    Pack type
    Pk
    Availability
    Price
  • APOSPC99020-500MG
    PC99020-500MG
    Bottle
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 3 380,00 Kč
    3 380,00 Kč
    1 * 500 mg
       3 380,00 Kč
    Supplier Article Number: PC99020-500MG
    Local Article Number: APOSPC99020-500MG
  • APOSPC99020-1G
    PC99020-1G
    Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 4 730,00 Kč
    4 730,00 Kč
    1 * 1 g
       4 730,00 Kč
    Supplier Article Number: PC99020-1G
    Local Article Number: APOSPC99020-1G