D-(+)-Allose
APOSBIA0110-1GEA
0
CZK
APOSBIA0110-1G
APOSBIA0110-500MG
D-(+)-Allose
D-(+)-Allose
Vzorec:
C₆H₁₂O₆ MW: 180,1559 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00135833 CAS číslo: 2595-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...