Tisk…

Amantadine hydrochloride

Varování

APOSBIA4305-25GEA 0 CZK
APOSBIA4305-25G APOSBIA4305-500G APOSBIA4305-100G
Amantadine hydrochloride
Amantadine hydrochloride
Vzorec: C₁₀H₁₈ClN
MW: 187,71 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00074723
CAS číslo: 665-66-7
EINECS: 211-560-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR