Tisk…

4-Methylumbelliferyl-β-D-lactoside

Dodavatel: Apollo Scientific

APOSBIB1477-100MGEA 10350 CZK
APOSBIB1477-100MG APOSBIB1477-250MG
4-Methylumbelliferyl-β-D-lactoside
4-Methylumbelliferyl-β-D-lactoside

4-Methylumbelliferyl-beta-D-lactoside is used as a MU fluorogenic substrate and chiral reagent, used for beta-lactosidase yielding a blue fluorescent solution upon cleavage.

Vzorec: C₂₂H₂₈O₁₃
Storage Temperature: Ambient
CAS číslo: 84325-23-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR