Tisk…

3-Nitro-L-tyrosine

APOSBIB6250-10GEA 0 CZK
APOSBIB6250-10G APOSBIB6250-25G
3-Nitro-L-tyrosine
3-Nitro-L-tyrosine
Vzorec: C₉H₁₀N₂O₅
MW: 226,3 g/mol
Bod tání: 233…235 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00007123
CAS číslo: 621-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR