Tisk…

3,3'-Diaminobenzidine tetrahydrochloride hydrate

Nebezpečí

APOSBID2041-5GEA 0 CZK
APOSBID2041-5G
3,3'-Diaminobenzidine tetrahydrochloride hydrate
3,3'-Diaminobenzidine tetrahydrochloride hydrate

Substrate for peroxidase. Also can be used for determination of selenium.

Vzorec: C₁₂H₁₄N₄·4HCl·H₂O
MW: 360,11 g/mol (anhy)
Bod tání: 280 °C
Bod vzplanutí: 200 °C
Storage Temperature: Freezer
MDL Number: MFCD08273058
CAS číslo: 868272-85-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR