L(-)-Talose
APOSBIFS0117-100MGEA
0
CZK
APOSBIFS0117-100MG
APOSBIFS0117-50MG
L(-)-Talose
L(-)-Talose
Vzorec:
C₆H₁₂O₆ Storage Temperature: Ambient |
MDL Number:
MFCD00135850 CAS číslo: 23567-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...