Hesperidin
APOSBIK9009-250GEA
0
CZK
APOSBIK9009-250G
APOSBIK9009-100G
APOSBIK9009-25G
Hesperidin
Hesperidin
Vzorec:
C₂₈H₃₄O₁₅ MW: 610,56 g/mol Bod tání: 250…255 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075663 CAS číslo: 520-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...