Tisk…

Okadaic acid sodium salt ≥98%

Dodavatel: Apollo Scientific
Nebezpečí

APOSBIM1579-100UGEA 0 CZK
APOSBIM1579-100UG APOSBIM1579-1MG APOSBIM1579-50UG
Okadaic acid sodium salt ≥98%
Okadaic acid sodium salt

Potent inhibitor of protein phosphatases, especially the PP-1 and PP-2 classes, in numerous cell types Trends Biochem. Sci. 15: 98-102 (1990).

Vzorec: C₄₄H₆₇NaO₁₃
MW: 827,00 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210208
CAS číslo: 209266-80-8
UN: 3462
ADR: 6.1,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR