3,6,9-Trioxa-1,11-undecanediamine
|
Nebezpečí
|
APOSBIPG1055-250MGEA
0
CZK
APOSBIPG1055-250MG
3,6,9-Trioxa-1,11-undecanediamine
3,6,9-Trioxa-1,11-undecanediamine
Vzorec:
C₈H₂₀N₂O₃ MW: 192,2560 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00628164 CAS číslo: 929-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...