Aluminium antimonide 99,95% 6.35 mm & down
Dodavatel: Apollo Scientific
APOSIN1008-100GEA
6550
CZK
APOSIN1008-100G
APOSIN1008-25G
Aluminium antimonide 99,95% 6.35 mm & down
Aluminium antimonide
Vzorec:
AlSb MW: 148,73 g/mol Bod tání: 1050 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014154 CAS číslo: 25152-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...