Dysprosium(III) acetate tetrahydrate 99,9%
APOSIN1618-100GEA
0
CZK
APOSIN1618-100G
Dysprosium(III) acetate tetrahydrate 99,9%
Dysprosium(III) acetate tetrahydrate
Vzorec:
C₆H₉DyO₆·4H₂O MW: 411,68 g/mol Bod tání: 120 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013033 CAS číslo: 15280-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...