Nickel(IV) sulphide 99%
|
Nebezpečí
|
APOSIN2716-5GEA
0
CZK
APOSIN2716-5G
Nickel(IV) sulphide 99%
Nickel(IV) sulphide
Vzorec:
NiS₂ MW: 122,82 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016267 CAS číslo: 12035-51-7 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...