Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium 99,9%
APOSIN7658-10GEA
0
CZK
APOSIN7658-10G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium 99,9%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium
Vzorec:
C₂₂H₃₈O₄Sr Bod tání: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064758 CAS číslo: 36830-74-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...