Tisk…

2-Naphthalen-2-ylpiperazine

Dodavatel: Apollo Scientific

APOSOR01725-50MGEA 1650 CZK
APOSOR01725-50MG APOSOR01725-500MG APOSOR01725-1G
2-Naphthalen-2-ylpiperazine
2-Naphthalen-2-ylpiperazine
Vzorec: C₁₄H₁₆N₂
MW: 212,30
Storage Temperature: Ambient
MDL Number: MFCD05864679
CAS číslo: 904816-32-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR