(±)-2-Piperazinecarboxylic acid dihydrochloride 98%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR0237-25GEA
2370
CZK
APOSOR0237-25G
(±)-2-Piperazinecarboxylic acid dihydrochloride 98%
(±)-2-Piperazinecarboxylic acid dihydrochloride
Vzorec:
C₅H₁₂Cl₂N₂O₂ MW: 203,07 g/mol Bod tání: 266…269 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044803 CAS číslo: 3022-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...