4-Acetamidothiophenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR0782-1GEA
2280
CZK
APOSOR0782-1G
APOSOR0782-5G
APOSOR0782-25G
4-Acetamidothiophenol 95%
4-Acetamidothiophenol
Vzorec:
C₈H₉NOS MW: 167,2282 g/mol Bod tání: 147…153 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004848 CAS číslo: 1126-81-4 EINECS: 214-427-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...