Pentachlorothiophenol
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR0794-25GEA
0
CZK
APOSOR0794-25G
APOSOR0794-5G
Pentachlorothiophenol
Pentachlorothiophenol
Vzorec:
C₆HCl₅S Bod tání: 223…227 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00059146 CAS číslo: 133-49-3 EINECS: 205-107-8 UN: 3335 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...