Tisk…

4-(2-Aminoethyl)tetrahydropyran 96%

Dodavatel: Apollo Scientific

Varování

APOSOR10327-1GEA 7670 CZK
APOSOR10327-1G APOSOR10327-5G
4-(2-Aminoethyl)tetrahydropyran 96%
4-(2-Aminoethyl)tetrahydropyran
Vzorec: C₇H₁₅NO
MW: 129,2001 g/mol
Bod varu: 88…89 °C/13mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD02179433
CAS číslo: 65412-03-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR