4-(Aminomethyl)tetrahydropyran hydrochloride 98%
|
Varování
|
APOSOR10334-100GEA
0
CZK
APOSOR10334-100G
APOSOR10334-5G
APOSOR10334-25G
4-(Aminomethyl)tetrahydropyran hydrochloride 98%
4-(Aminomethyl)tetrahydropyran hydrochloride
Vzorec:
C₆H₁₄ClNO MW: 151,6300 g/mol Bod tání: 180…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411928 CAS číslo: 389621-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...