2-(Benzyloxy)phenylboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR10360-25GEA
7540
CZK
APOSOR10360-25G
APOSOR10360-5G
2-(Benzyloxy)phenylboronic acid
2-(Benzyloxy)phenylboronic acid
Vzorec:
C₁₃H₁₃BO₃ MW: 228,06 Bod tání: 105…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01632206 CAS číslo: 190661-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...