4-(tert-Butylcarbamoyl)phenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR10398-5GEA
5630
CZK
APOSOR10398-5G
APOSOR10398-250MG
APOSOR10398-1G
4-(tert-Butylcarbamoyl)phenylboronic acid 97%
4-(tert-Butylcarbamoyl)phenylboronic acid
Vzorec:
C₁₁H₁₆BNO₃ MW: 221,06 g/mol Bod tání: 230…246 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788413 CAS číslo: 850568-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...