4-Carboxy-2-nitrophenylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
APOSOR10401-1GEA
748
CZK
APOSOR10401-1G
4-Carboxy-2-nitrophenylboronic acid 97%
4-Carboxy-2-nitrophenylboronic acid
Vzorec:
C₇H₆BNO₆ MW: 210,94 Bod tání: 265…268 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00448780 CAS číslo: 85107-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...