Tisk…

4-Isopropylphenylboronic acid

Dodavatel: Apollo Scientific
Varování

APOSOR10524-5GEA 1990 CZK
APOSOR10524-5G APOSOR10524-25G
4-Isopropylphenylboronic acid
4-Isopropylphenylboronic acid
Vzorec: C₉H₁₃BO₂
MW: 164,01 g/mol
Bod tání: 140…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074614
CAS číslo: 16152-51-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR