Tisk…

tert-Butyl (piperidin-4-ylmethyl)carbamate 95%

Nebezpečí

APOSOR10830-25GEA 8440 CZK
APOSOR10830-25G APOSOR10830-5G APOSOR10830-1G
tert-Butyl (piperidin-4-ylmethyl)carbamate 95%
tert-Butyl (piperidin-4-ylmethyl)carbamate
Vzorec: C₁₁H₂₂N₂O₂
MW: 214,31 g/mol
Bod tání: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631214
CAS číslo: 135632-53-0
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR