5-(3-Bromophenyl)isoxazole
APOSOR11-1GEA
0
CZK
APOSOR11-1G
5-(3-Bromophenyl)isoxazole
5-(3-Bromophenyl)isoxazole
Vzorec:
C₉H₆BrNO MW: 224,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02183531 CAS číslo: 7064-33-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...