4-(4-Formylphenoxy)benzonitrile
Dodavatel: Apollo Scientific
APOSOR110983-1GEA
2490
CZK
APOSOR110983-1G
4-(4-Formylphenoxy)benzonitrile
4-(4-Formylphenoxy)benzonitrile
Vzorec:
C₁₄H₉NO₂ MW: 223,2300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08276813 CAS číslo: 90178-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...